Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh02845434_m1
See other ZNF7 GE Assays ›
Gene Symbol:
ZNF7
Gene Name:
zinc finger protein 7
Gene Aliases:
-
Chromosome Location:
Chr.8: 145144786 - 145160013 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh02845434_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ZNF7
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001266571.2
NP_001253500.1
149
XM_015146220.2
XP_015001706.2
726
149
XM_015146222.2
XP_015001708.2
400
149
XM_015146223.2
XP_015001709.2
825
149
XM_015146224.2
XP_015001710.2
1134
149
XM_015146225.2
XP_015001711.2
479
149
XM_015146227.2
XP_015001713.2
763
149
XM_015146228.2
XP_015001714.2
317
149
XM_015146229.2
XP_015001715.2
445
149
XM_015146230.2
XP_015001716.2
757
149
XM_015146231.2
XP_015001717.2
397
149
XM_015146232.2
XP_015001718.2
309
149
XM_015146233.2
XP_015001719.2
758
149
XM_015146234.2
XP_015001720.2
306
149
XM_015146235.2
XP_015001721.2
416
149
XM_015146236.2
XP_015001722.2
437
149
XM_015146238.2
XP_015001724.2
664
149
XM_015146239.2
XP_015001725.2
380
149
XM_015146240.2
XP_015001726.2
661
149
XM_015146242.2
XP_015001728.2
558
149
XM_015146243.2
XP_015001729.2
731
149
XM_015146244.2
XP_015001730.2
256
149
XM_028852265.1
XP_028708098.1
493
149
XM_028852267.1
XP_028708100.1
775
149
XM_028852268.1
XP_028708101.1
773
149
XM_028852269.1
XP_028708102.1
525
149
GenBank mRNA
JU323957.1
-
149
JU472655.1
-
149
JV045382.1
-
149
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015146242.2, XM_015146243.2, XM_015146244.2, XM_028852265.1, XM_028852269.1 transcripts.
Set Membership:
Amplicon length greater than or equal to 101
3' Most
Recommended Assay
Macaca mulatta
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-bsnw9:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0