Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015109773.2 | XP_014965259.1 | 465 | 121 | ||
XM_028830248.1 | XP_028686081.1 | 477 | 121 | |||
GenBank mRNA | JU493010.1 | - | 243 | 121 | ||
JV070639.1 | - | 283 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |