Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194124.2 | NP_001181053.2 | 659 | 108 | ||
XM_028844987.1 | XP_028700820.1 | 659 | 108 | |||
GenBank mRNA | JU336283.1 | - | 640 | 108 | ||
JU336284.1 | - | 640 | 108 | |||
JU336285.1 | - | 640 | 108 | |||
JU472805.1 | - | 635 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |