Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085459.4 | XP_001085459.3 | 1379 | 130 | ||
XM_015123477.2 | XP_014978963.2 | 942 | 130 | |||
XM_015123478.2 | XP_014978964.2 | 1032 | 130 | |||
XM_015123480.2 | XP_014978966.2 | 911 | 130 | |||
XM_015123483.2 | XP_014978969.2 | 1342 | 130 | |||
XM_028839928.1 | XP_028695761.1 | 1215 | 130 | |||
GenBank mRNA | JV664417.1 | - | 1051 | 130 | ||
JV699930.1 | - | 1029 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |