Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257649.1 | NP_001244578.1 | 903 | 106 | ||
XM_015143946.2 | XP_014999432.1 | 1309 | 106 | |||
XM_015143947.2 | XP_014999433.1 | 733 | 106 | |||
XM_015143948.2 | XP_014999434.1 | 841 | 106 | |||
XM_015143949.2 | XP_014999435.1 | 754 | 106 | |||
GenBank mRNA | JU321129.1 | - | 903 | 106 | ||
JV043369.1 | - | 905 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |