Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001115576.4 | XP_001115576.1 | 482 | 128 | ||
XM_015122974.2 | XP_014978460.1 | 548 | 128 | |||
XM_015122993.2 | XP_014978479.2 | 545 | 128 | |||
XM_015123008.2 | XP_014978494.1 | 547 | 128 | |||
XM_015123025.2 | XP_014978511.1 | 378 | 128 | |||
XM_028850751.1 | XP_028706584.1 | 551 | 128 | |||
XM_028850762.1 | XP_028706595.1 | 443 | 128 | |||
XM_028850766.1 | XP_028706599.1 | 378 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |