Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AJUBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104222.4 | XP_001104222.1 | 2289 | 101 | ||
GenBank mRNA | JV073934.1 | - | 819 | 101 | ||
JV663941.1 | - | 1083 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |