Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139710.2 | XP_014995196.2 | 1019 | 112 | ||
XM_028849326.1 | XP_028705159.1 | 1019 | 112 | |||
GenBank mRNA | JV052977.1 | - | 106 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |