Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133199.2 | XP_014988685.1 | 1167 | 74 | ||
XM_028845520.1 | XP_028701353.1 | 2001 | 74 | |||
XM_028845521.1 | XP_028701354.1 | 1167 | 74 |