Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091313.4 | XP_001091313.1 | 2323 | 131 | ||
XM_028841937.1 | XP_028697770.1 | 2226 | 131 | |||
XM_028841938.1 | XP_028697771.1 | 2134 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |