Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001114413.4 | XP_001114413.3 | 1216 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |