Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028837038.1 | XP_028692871.1 | 1106 | 78 | ||
XM_028837039.1 | XP_028692872.1 | 1106 | 78 | |||
GenBank mRNA | JU326626.1 | - | 790 | 78 | ||
JU485939.1 | - | 822 | 78 | |||
JV650494.1 | - | 1053 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |