Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265938.2 | NP_001252867.1 | 487 | 122 | ||
GenBank mRNA | JU320644.1 | - | 487 | 122 | ||
JV529645.1 | - | 350 | 122 | |||
JV669313.1 | - | 517 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |