Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122790.2 | XP_014978276.1 | 1846 | 91 | ||
XM_015122797.2 | XP_014978283.1 | 2190 | 91 | |||
XM_015122805.2 | XP_014978291.1 | 1893 | 91 | |||
XM_015122815.2 | XP_014978301.1 | 2126 | 91 | |||
XM_015122831.2 | XP_014978317.1 | 1956 | 91 | |||
XM_015122840.2 | XP_014978326.1 | 2189 | 91 | |||
XM_015122847.2 | XP_014978333.1 | 1893 | 91 | |||
XM_015122856.2 | XP_014978342.1 | 2126 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |