Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265893.1 | NP_001252822.1 | 1276 | 117 | ||
XM_015116917.2 | XP_014972403.1 | 1540 | 117 | |||
XM_015116918.2 | XP_014972404.1 | 1345 | 117 | |||
XM_028834257.1 | XP_028690090.1 | 1299 | 117 | |||
GenBank mRNA | JU472522.1 | - | 1276 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |