Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100646.4 | XP_001100646.3 | 2527 | 87 | ||
GenBank mRNA | JU336258.1 | - | 1440 | 87 | ||
JV048170.1 | - | 1487 | 87 | |||
JV627873.1 | - | 1403 | 87 | |||
JV660585.1 | - | 1488 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |