Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151956.2 | XP_015007442.1 | 191 | 97 | ||
XM_015151957.2 | XP_015007443.1 | 184 | 97 | |||
XM_015151960.2 | XP_015007446.1 | 184 | 97 | |||
XM_028828955.1 | XP_028684788.1 | 186 | 97 | |||
GenBank mRNA | JU332442.1 | - | 161 | 97 | ||
JU473908.1 | - | 82 | 97 | |||
JV045863.1 | - | 149 | 97 | |||
JV601081.1 | - | 156 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |