Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD2BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260696.1 | NP_001247625.1 | 80 | 117 | ||
GenBank mRNA | JU332503.1 | - | 77 | 117 | ||
JV045903.1 | - | 80 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |