Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194304.2 | NP_001181233.1 | 582 | 68 | ||
XM_015133907.2 | XP_014989393.1 | 547 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |