Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260665.1 | NP_001247594.1 | 315 | 154 | ||
GenBank mRNA | JU320037.1 | - | 339 | 154 | ||
JU471482.1 | - | 315 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |