Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260569.2 | NP_001247498.1 | 546 | 89 | ||
GenBank mRNA | JU320529.1 | - | 912 | 89 | ||
JU320530.1 | - | 649 | 89 | |||
JV043744.1 | - | 546 | 89 | |||
JV703962.1 | - | 1061 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |