Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015147516.2 | XP_015003002.1 | 1408 | 117 | ||
XM_015147517.2 | XP_015003003.1 | 695 | 117 | |||
GenBank mRNA | JU326950.1 | - | 1070 | 117 | ||
JV710793.1 | - | 1091 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |