Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261795.2 | NP_001248724.1 | 1270 | 124 | ||
XM_015135555.2 | XP_014991041.1 | 1336 | 124 | |||
GenBank mRNA | JU321479.1 | - | 1407 | 124 | ||
JU473822.1 | - | 1258 | 124 | |||
JV045677.1 | - | 1272 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |