Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh02864756_m1
See other POLL GE Assays ›
Gene Symbol:
POLL
Gene Name:
DNA polymerase lambda
Gene Aliases:
EGK_19997
Chromosome Location:
Chr.9: 102054715 - 102075556 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh02864756_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: POLL
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001266906.2
NP_001253835.1
1231
90
XM_015147969.2
XP_015003455.1
1365
90
XM_015147970.2
XP_015003456.1
1405
90
XM_015147971.2
XP_015003457.1
1402
90
XM_015147972.2
XP_015003458.1
1455
90
XM_015147973.2
XP_015003459.1
1403
90
XM_015147974.2
XP_015003460.1
1265
90
XM_015147975.2
XP_015003461.1
1191
90
XM_015147978.2
XP_015003464.1
1089
90
XM_015147980.2
XP_015003466.1
629
90
XM_015147981.2
XP_015003467.1
595
90
XM_015147982.2
XP_015003468.1
605
90
XM_028853470.1
XP_028709303.1
1277
90
XM_028853471.1
XP_028709304.1
1687
90
XM_028853472.1
XP_028709305.1
1228
90
XM_028853473.1
XP_028709306.1
1402
90
XM_028853475.1
XP_028709308.1
1402
90
XM_028853476.1
XP_028709309.1
1126
90
XM_028853480.1
XP_028709313.1
626
90
XM_028853481.1
XP_028709314.1
498
90
XM_028853482.1
XP_028709315.1
901
90
GenBank mRNA
JU334040.1
-
1245
90
JV046782.1
-
1232
90
JV046783.1
-
1232
90
JV637757.1
-
1541
90
Back To Top
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership:
Amplicon length between 86 and 100
3' Most
Macaca mulatta
Panther Classification:
Molecular Function -
DNA-directed DNA polymerase
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-4vcgr:80/10.239.160.220:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0