Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260925.1 | NP_001247854.1 | 492 | 122 | ||
GenBank mRNA | JU334159.1 | - | 492 | 122 | ||
JV579572.1 | - | 482 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |