Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258195.1 | NP_001245124.1 | 659 | 114 | ||
XM_028837742.1 | XP_028693575.1 | 651 | 114 | |||
GenBank mRNA | JU328027.1 | - | 445 | 114 | ||
JU475473.1 | - | 659 | 114 | |||
JV636131.1 | - | 541 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |