Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100732.4 | XP_001100732.2 | 497 | 165 | ||
XM_015124042.2 | XP_014979528.2 | 239 | 165 | |||
XM_028850415.1 | XP_028706248.1 | 503 | 165 | |||
GenBank mRNA | JU334889.1 | - | 165 | |||
JU334890.1 | - | 165 | ||||
JU334891.1 | - | 165 | ||||
JU475899.1 | - | 165 | ||||
JV047300.1 | - | 454 | 165 | |||
JV047301.1 | - | 454 | 165 | |||
JV636208.1 | - | 411 | 165 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |