Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194605.2 | NP_001181534.1 | 136 | 100 | ||
XM_015126044.2 | XP_014981530.1 | 459 | 100 | |||
GenBank mRNA | JU474980.1 | - | 113 | 100 | ||
JV611501.1 | - | 132 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |