Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257462.2 | NP_001244391.1 | 2175 | 89 | ||
XM_015141871.2 | XP_014997357.1 | 2540 | 89 | |||
XM_015141872.2 | XP_014997358.1 | 2408 | 89 | |||
XM_015141873.2 | XP_014997359.1 | 2551 | 89 | |||
XM_015141874.2 | XP_014997360.1 | 2459 | 89 | |||
XM_015141877.2 | XP_014997363.1 | 2416 | 89 | |||
XM_028850301.1 | XP_028706134.1 | 2199 | 89 | |||
XM_028850302.1 | XP_028706135.1 | 2196 | 89 | |||
XM_028850303.1 | XP_028706136.1 | 2165 | 89 | |||
XM_028850304.1 | XP_028706137.1 | 2309 | 89 | |||
GenBank mRNA | JU321091.1 | - | 2175 | 89 | ||
JU333471.1 | - | 2166 | 89 | |||
JU471184.1 | - | 2178 | 89 | |||
JU474747.1 | - | 2169 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |