Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JADE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015140931.2 | XP_014996417.1 | 699 | 103 | ||
XM_015140932.2 | XP_014996418.1 | 700 | 103 | |||
XM_015140933.2 | XP_014996419.1 | 702 | 103 | |||
XM_015140934.2 | XP_014996420.1 | 703 | 103 | |||
XM_015140935.2 | XP_014996421.1 | 831 | 103 | |||
XM_015140936.2 | XP_014996422.1 | 723 | 103 | |||
XM_015140937.2 | XP_014996423.1 | 556 | 103 | |||
XM_015140939.2 | XP_014996425.1 | 1064 | 103 | |||
XM_015140940.2 | XP_014996426.1 | 592 | 103 | |||
XM_015140941.2 | XP_014996427.1 | 831 | 103 | |||
GenBank mRNA | JU333651.1 | - | 569 | 103 | ||
JU474902.1 | - | 488 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |