Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260862.1 | NP_001247791.1 | 729 | 146 | ||
XM_015141196.2 | XP_014996682.1 | 837 | 146 | |||
GenBank mRNA | JU472300.1 | - | 729 | 146 | ||
JU475109.1 | - | 729 | 146 | |||
JV057555.1 | - | 583 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |