Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOSIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194679.2 | NP_001181608.2 | 644 | 131 | ||
XM_015124396.2 | XP_014979882.2 | 658 | 131 | |||
GenBank mRNA | JU322037.1 | - | 540 | 131 | ||
JU471600.1 | - | 544 | 131 | |||
JV452578.1 | - | 535 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |