Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266056.1 | NP_001252985.1 | 1034 | 111 | ||
GenBank mRNA | JU322099.1 | - | 1034 | 111 | ||
JV549051.1 | - | 660 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |