Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TESC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257539.1 | NP_001244468.1 | 213 | 76 | ||
GenBank mRNA | JU320871.1 | - | 213 | 76 | ||
JV044375.1 | - | 266 | 76 |