Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261104.2 | NP_001248033.1 | 502 | 105 | ||
XM_028852156.1 | XP_028707989.1 | 1428 | 105 | |||
GenBank mRNA | JU320971.1 | - | 395 | 105 | ||
JU473809.1 | - | 487 | 105 | |||
JV045168.1 | - | 502 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta Inventoried |