Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092640.4 | XP_001092640.1 | 241 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |