Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL36G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090647.4 | XP_001090647.1 | 3813 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |