Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATAD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015109702.2 | XP_014965188.1 | 1893 | 60 | ||
XM_015109710.2 | XP_014965196.1 | 1890 | 60 | |||
GenBank mRNA | JU320721.1 | - | 1767 | 60 | ||
JU473249.1 | - | 1750 | 60 | |||
JV044414.1 | - | 1773 | 60 | |||
JV567988.1 | - | 1774 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |