Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257565.1 | NP_001244494.1 | 1255 | 106 | ||
GenBank mRNA | JU335621.1 | - | 1255 | 106 | ||
JU476535.1 | - | 1274 | 106 | |||
JV527811.1 | - | 1285 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |