Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257653.1 | NP_001244582.1 | 548 | 68 | ||
GenBank mRNA | JU335656.1 | - | 548 | 68 | ||
JU476557.1 | - | 438 | 68 | |||
JV565626.1 | - | 627 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |