Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257463.1 | NP_001244392.1 | 637 | 106 | ||
GenBank mRNA | JU336155.1 | - | 637 | 106 | ||
JU477012.1 | - | 554 | 106 | |||
JV048101.1 | - | 636 | 106 | |||
JV592263.1 | - | 639 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |