Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRINP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265618.1 | NP_001252547.1 | 981 | 97 | ||
XM_028843587.1 | XP_028699420.1 | 983 | 97 | |||
XM_028843589.1 | XP_028699422.1 | 1139 | 97 | |||
XM_028843593.1 | XP_028699426.1 | 817 | 97 | |||
GenBank mRNA | JU321881.1 | - | 981 | 97 | ||
JV667437.1 | - | 981 | 97 |