Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028835225.1 | XP_028691058.1 | 1295 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |