Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001116097.4 | XP_001116097.2 | 1065 | 96 | ||
XM_015149479.2 | XP_015004965.1 | 444 | 96 | |||
GenBank mRNA | JV658479.1 | - | 288 | 96 |