Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194202.2 | NP_001181131.2 | 1010 | 92 | ||
XM_015125570.2 | XP_014981056.2 | 1320 | 92 | |||
XM_015125571.2 | XP_014981057.2 | 1333 | 92 | |||
XM_028840519.1 | XP_028696352.1 | 1320 | 92 | |||
GenBank mRNA | JU323753.1 | - | 1246 | 92 | ||
JU472906.1 | - | 1134 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |