Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP4K2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261154.1 | NP_001248083.1 | 777 | 149 | ||
GenBank mRNA | JU336433.1 | - | 769 | 149 | ||
JU477249.1 | - | 746 | 149 | |||
JV048280.1 | - | 777 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |