Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047138.1 | NP_001040603.1 | 373 | 150 | ||
GenBank mRNA | JV490465.1 | - | 387 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |