Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015135046.2 | XP_014990532.1 | 2469 | 79 | ||
GenBank mRNA | JU326136.1 | - | 2358 | 79 | ||
JV724219.1 | - | 2270 | 79 |