Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266728.2 | NP_001253657.1 | 1171 | 124 | ||
GenBank mRNA | JU322213.1 | - | 1158 | 124 | ||
JU471161.1 | - | 1171 | 124 | |||
JV638636.1 | - | 1180 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |