Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266132.1 | NP_001253061.1 | 55 | 114 | ||
XM_015116635.2 | XP_014972121.1 | 127 | 114 | |||
GenBank mRNA | JU336544.1 | - | 77 | 114 | ||
JU471574.1 | - | 64 | 114 | |||
JU477360.1 | - | 64 | 114 | |||
JV048340.1 | - | 55 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |